missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(+)-Camphor-10-sulfonic acid, 98+%(dry wt.), water <2%

Product Code. 11473247
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473247 500 g 500g
11453247 25 g 25g
11463247 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473247 Supplier Thermo Scientific Alfa Aesar Supplier No. A16179.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3144-16-9 | C10H15O4S | 231.29 g/mol

(1S)-(+)-Camphor-10-sulfonic acid is used as stabilizer. (+)-(1S)-camphor-10-sulfonic acid being a very effective resolving agent. . Chiral polyaniline(PANI) nanofibers were synthesized via facilely potentiostatic electropolymerization method without template in the presence of(1S)-(+)camphor-10-sulfonic acid(D-CSA) or(1R)-(-)camphor-10-sulfonic acid(L-CSA) as the dopant. Synthesis of QUATs derived from (1S)-(+)camphor-10-sulfonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S)-(+)-Camphor-10-sulfonic acid is used as stabilizer. (+)-(1S)-camphor-10-sulfonic acid being a very effective resolving agent. . Chiral polyaniline(PANI) nanofibers were synthesized via facilely potentiostatic electropolymerization method without template in the presence of(1S)-(+)camphor-10-sulfonic acid(D-CSA) or(1R)-(-)camphor-10-sulfonic acid(L-CSA) as the dopant. Synthesis of QUATs derived from (1S)-(+)camphor-10-sulfonic acid.

Solubility
Exhibit moderate solubility in HCCl.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases. Keep under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3144-16-9
Molecular Formula C10H15O4S
Molecular Weight (g/mol) 231.29
MDL Number MFCD00064157,MFCD00074827
InChI Key MIOPJNTWMNEORI-XVKPBYJWSA-M
Synonym 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid
PubChem CID 65617
ChEBI CHEBI:55403
IUPAC Name [(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
SMILES CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2

Specifications

Melting Point 196°C to 200°C (decomposition)
Quantity 500 g
UN Number UN3261
Beilstein 2809675
Sensitivity Hygroscopic
Merck Index 14,1734
Solubility Information Exhibit moderate solubility in HCCl.
Optical Rotation +20° (c=2 in Water)
Formula Weight 232.3
Percent Purity ≥98%
Assay (dry wt.), water <2%
Chemical Name or Material (1S)-(+)-Camphor-10-sulfonic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.