missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX ADP HunterTM Plus
Click to view available options
Quantity:
100 mL (10,000 dp, 384-well)
Unit Size:
1 set
Description
The ADP HunterTM Plus kit is a detection kit for the measurement of ADP produced as a product of kinase activity. It is used to detect substrate-induced activity of purified or semi-purified kinases. ADP produced by the kinase reacts with intermediate factors to produce a fluorescent dye precursor, which generates signal when combined with peroxide and peroxidase. This detection kit contains reagents for 25 plates (384-well).
Specifications
Specifications
| Assay Entry | Kinase activity |
| For Use With (Application) | Detection Kits |
| Quantity | 100 mL (10,000 dp, 384-well) |
| Target Entry | Any kinase, ATPase, GTPase or UTPase |
| Target | Any kinase, ATPase, GTPase or UTPase |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction