missing translation for 'onlineSavingsMsg'
Learn More

2-(1,1,1-Tributylstannyl)pyridine, Tech., Thermo Scientific™

Product Code. 10762493
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10762493 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10762493 Supplier Thermo Scientific Maybridge Supplier No. KM05480DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 17997-47-6
Density 1.15g/mL
Packaging Amber Glass Bottle
Molecular Formula C17H31NSn
MDL Number MFCD00052052
Quantity 1 g
Synonym 2-tributylstannyl pyridine, 2-tri-n-butylstannylpyridine, 2-tributylstannylpyridine, tributyl 2-pyridyl tin, 2-pyridyltributyltin, 2-1,1,1-tributylstannyl pyridine, 2-pyridyltri-n-butyltin, 2-tributylstannyl-pyridine, tri-n-butyl 2-pyridyl tin
InChI Key GYUURHMITDQTRU-UHFFFAOYSA-N
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CC=N1
IUPAC Name tributyl(pyridin-2-yl)stannane
Molecular Weight (g/mol) 368.152
PubChem CID 607784
Formula Weight 368.15
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-(1, 1, 1-Tributylstannyl)pyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.