missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazole-4-carbaldehyde, 95%, Thermo Scientific™

Product Code. 10552842
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10552842 250 mg 250mg
10049141 1 g 1g
10732027 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10552842 Supplier Thermo Scientific Maybridge Supplier No. CC09104CB
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 5170-68-3
Molecular Formula C7H4N2OS
Molecular Weight (g/mol) 164.182
MDL Number MFCD02681902
InChI Key ANVJARPTPIVPRC-UHFFFAOYSA-N
Synonym benzo c 1,2,5 thiadiazole-4-carbaldehyde, 2,1,3-benzothiadiazole-4-carboxaldehyde, tos-bb-1233, 2,1,3-benzothiadiazol-4-aldehyde, 2,1,3-benzothiadiazole-4-aldehyde, 2,1,3-benzothiadiazol-4-carboxaldehyde, benzo c 1,2,5-thiadiazole-4-carbaldehyde, 2,1,3-benzothiadiazole-4-carbaldehyde
PubChem CID 2776283
IUPAC Name 2,1,3-benzothiadiazole-4-carbaldehyde
SMILES C1=CC2=NSN=C2C(=C1)C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 164.19
Percent Purity 95%
Chemical Name or Material 2, 1, 3-Benzothiadiazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.