missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10782283
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10782283 1 g Amber Glass Bottle 1g
10772283 250 mg Amber Glass Bottle 250mg
10462434 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10782283 Supplier Thermo Scientific Maybridge Supplier No. CC09101DA
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 3529-57-5
Molecular Formula C7H4N2O2S
Molecular Weight (g/mol) 180.181
MDL Number MFCD05664666
InChI Key ZGDGZMOKXTUMEV-UHFFFAOYSA-N
Synonym benzo c 1,2,5 thiadiazole-4-carboxylic acid, 4-carboxybenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole-4-carboxylicacid, 1,2,3-benzothiadiazole-4-carboxylic acid, benzo 1,2,5 thiadiazole-4-carboxylic acid, benzo c 1,2,5-thiadiazole-4-carboxylic acid, 2,1,3-benzothiadiazole-2-siv-4-carboxylic acid, 2??,1,3-benzothiadiazole-4-carboxylic acid
PubChem CID 2795202
IUPAC Name 2,1,3-benzothiadiazole-4-carboxylic acid
SMILES C1=CC2=NSN=C2C(=C1)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 180.19
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzothiadiazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.