missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazole-4-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10344472
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10344472 1 g 1g
10214934 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10344472 Supplier Thermo Scientific Maybridge Supplier No. CC09103DA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Identificadores de productos químicos

CAS 73713-79-8
Molecular Formula C6H3ClN2O2S2
Molecular Weight (g/mol) 234.67
MDL Number MFCD00068049
InChI Key CXAICGCTHOWKPP-UHFFFAOYSA-N
Synonym benzo c 1,2,5 thiadiazole-4-sulfonyl chloride, benzo 2,1,3 thiadiazole-4-sulfonyl chloride, 2,1,3-benzothiadiazole-4-sulphonyl chloride, 2,1,3-benzothiadiazol-4-sulfonyl chloride, 4-chlorosulfonyl-2,1,3-benzothiadiazole, 2,1,3-benzothiadiazole, 4-chlorosulfonyl, 2,1,3-benzothiadiazole-4-sulfonylchloride, benzo-2,1,3-thiadiazole-4-sulfonyl chloride, 2??,1,3-benzothiadiazole-4-sulfonyl chloride, benzo-2,1,3-thiadiazole-4-sulphonyl chloride
PubChem CID 2735459
IUPAC Name 2,1,3-benzothiadiazole-4-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=CC2=NSN=C12

Especificaciones

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 234.69
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzothiadiazole-4-sulfonyl chloride
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.