missing translation for 'onlineSavingsMsg'
Learn More

2-(1,3-Dioxan-2-yl)ethylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15485425
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485425 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15485425 Supplier Thermo Scientific Alfa Aesar Supplier No. H58912.AD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 307531-82-4
Density 0.982
Molecular Formula C6H11BrO2Zn
MDL Number MFCD02260128,MFCD02260128
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Synonym 1,3-dioxan-2-ylethyl zinc bromide
InChI Key ALTXXOLEYMBUCK-UHFFFAOYSA-M
SMILES [Zn++].[Br-].[CH2-]CC1OCCCO1
IUPAC Name 2-ethyl-1,3-dioxane;zinc;bromide
Molecular Weight (g/mol) 260.44
PubChem CID 131668528
Formula Weight 260.44
Chemical Name or Material 2-(1,3-Dioxan-2-yl)ethylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.