missing translation for 'onlineSavingsMsg'
Learn More

2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester, 97%

Product Code. 11392009
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392009 5 g 5g
11382009 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392009 Supplier Thermo Scientific Alfa Aesar Supplier No. H26071.06
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1073354-07-0 | C11H21BO4 | 228.095 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073354-07-0
Molecular Formula C11H21BO4
Molecular Weight (g/mol) 228.095
MDL Number MFCD03788722
InChI Key DNBRLKJRBDIKOO-UHFFFAOYSA-N
Synonym 2-2-1,3-dioxolan-2-yl ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1,3-dioxolan-2-yl ethylboronic acid pinacol ester, 2-1,3-dioxolan-2-yl-1-ethylboronic acid pinacol ester
PubChem CID 46739008
IUPAC Name 2-[2-(1,3-dioxolan-2-yl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCC2OCCO2

Specifications

Boiling Point 100°C (1.5 mmHg)
Quantity 5 g
Formula Weight 228.1
Percent Purity 97%
Chemical Name or Material 2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.