missing translation for 'onlineSavingsMsg'
Learn More

2-(1,3-Dioxolan-2-yl)ethylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 15495425
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495425 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15495425 Supplier Thermo Scientific Alfa Aesar Supplier No. H58916.AD
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 307531-83-5 | C5H9BrO2Zn | 246.409 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in pharmaceutical industry.

Solubility
Reacts with water.

Notes
Handle under dry protective gas. Keep away from strong oxidizing agents, heat, acid chlorides, bases and acids.
TRUSTED_SUSTAINABILITY

Specifications

CAS 307531-83-5
Density 0.984
Molecular Formula C5H9BrO2Zn
MDL Number MFCD02260129
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Synonym zinc 2+ ion 2-1,3-dioxolan-2-yl ethanide bromide
Solubility Information Reacts with water.
InChI Key FBIHMCAQTQOHLI-UHFFFAOYSA-M
SMILES [CH2-]CC1OCCO1.[Zn+2].[Br-]
IUPAC Name zinc;2-ethyl-1,3-dioxolane;bromide
Molecular Weight (g/mol) 246.409
PubChem CID 57377022
Formula Weight 246.41
Chemical Name or Material 2-(1,3-Dioxolan-2-yl)ethylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.