missing translation for 'onlineSavingsMsg'
Learn More

2-(1,4-Diazepan-1-yl)ethan-1-ol, 97%, Thermo Scientific™

Product Code. 10772073
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10772073 1 g Amber Glass Bottle 1g
10334702 250 mg Amber Glass Bottle 250mg
10244924 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10772073 Supplier Thermo Scientific Maybridge Supplier No. AW00182DA
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 53427-65-9
Molecular Formula C7H16N2O
Molecular Weight (g/mol) 144.218
MDL Number MFCD01565842
InChI Key IXKNVDAHLCFGCV-UHFFFAOYSA-N
Synonym 2-1,4-diazepan-1-yl ethanol, 2-1,4-diazepan-1-yl ethan-1-ol, 1h-1,4-diazepine-1-ethanol, hexahydro, 2-1,4-diazaperhydroepinyl ethan-1-ol, 1-homopiperazineethanol, 1-2-hydroxyethyl homopiperazine, 2-1,4 diazepan-1-yl-ethanol, 1-2-hydroxyethyl-homopiperazine, 1-2-hydroxyethyl homo-piperazine, 1-2-hydroxyethyl-1,4-diazepane
PubChem CID 2736550
IUPAC Name 2-(1,4-diazepan-1-yl)ethanol
SMILES C1CNCCN(C1)CCO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 144.22
Percent Purity 97%
Chemical Name or Material 2-(1, 4-Diazepan-1-yl)ethan-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.