missing translation for 'onlineSavingsMsg'
Learn More

2-(1,4-Oxazinan-2-ylmethyl)-1H-isoindole-1,3(2H)-dione, Thermo Scientific™

Product Code. 10171432
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10171432 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10171432 Supplier Thermo Scientific Maybridge Supplier No. TL01033DA
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 499771-20-9
Assay Percent Range 90%
Packaging Amber Glass Bottle
Molecular Formula C13H14N2O3
Quantity 1 g
Synonym 2-morpholin-2-ylmethyl isoindoline-1,3-dione, 2-morpholin-2-ylmethyl isoindole-1,3-dione, 2-1,4-oxazinan-2-ylmethyl-1h-isoindole-1,3 2h-dione, 2-2-morpholinylmethyl isoindole-1,3-dione, 2-morpholin-2-ylmethyl benzo c azoline-1,3-dione, 2-morpholin-2-yl methyl-1h-isoindole-1,3 2h-dione, 2-morpholin-2-ylmethyl-2,3-dihydro-1h-isoindole-1,3-dione
InChI Key UXEHWYCZZGPXOH-UHFFFAOYSA-N
SMILES C1COC(CN1)CN2C(=O)C3=CC=CC=C3C2=O
IUPAC Name 2-(morpholin-2-ylmethyl)isoindole-1,3-dione
Molecular Weight (g/mol) 246.266
PubChem CID 2801580
Formula Weight 246.26
Percent Purity 90%
Chemical Name or Material 2-(1, 4-Oxazinan-2-ylmethyl)-1H-isoindole-1, 3(2H)-dione
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.