missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Adamantyl)propan-2-ol, 97%, Thermo Scientific™

Product Code. 10641426
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10641426 5 g 5g
10611303 250 mg 250mg
10475512 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10641426 Supplier Thermo Scientific Maybridge Supplier No. NRB01891DE
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 775-64-4
Molecular Formula C13H22O
Molecular Weight (g/mol) 194.32
MDL Number MFCD00167855
InChI Key WBKAUEBLTWRERU-UHFFFAOYSA-N
Synonym 2-adamantan-1-yl-propan-2-ol, 2-1-adamantyl propan-2-ol, 2-adamantan-1-yl propan-2-ol, 2-1-adamantyl-2-propanol, 2-adamantanylpropan-2-ol, pubchem14445, maybridge1_005546, 2-1-adamantly-2-propanol, 2-1-adamantyl-propan-2-ol
PubChem CID 300814
SMILES CC(C)(O)C12CC3CC(CC(C3)C1)C2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
IUPAC Name 2-(adamantan-1-yl)propan-2-ol
Formula Weight 194.32
Percent Purity 97%
Chemical Name or Material 2-(1-Adamantyl)propan-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.