missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Benzofuran-3-yl)ethyl 4-methylbenzenesulfonate, ≥97%, Thermo Scientific™

Product Code. 10623065
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10623065 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10623065 Supplier Thermo Scientific Maybridge Supplier No. CC40844DA
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 26278-25-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C17H16O4S
MDL Number MFCD08271906
Quantity 1 g
Synonym 2-1-benzofuran-3-yl ethyl 4-methylbenzenesulfonate, 3-benzofuranethanol,3-4-methylbenzenesulfonate, p-toluenesulfonic acid 2-benzofuran-3-yl ethyl ester, 2-1-benzofuran-3-yl ethyl 4-methylbenzene-1-sulfonate, 2-1-benzo b furan-3-yl ethyl-4-methylbenzenesulfonate
InChI Key OEJOFPWUQPRTDR-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCC1=COC2=CC=CC=C12
IUPAC Name 2-(1-benzofuran-3-yl)ethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol) 316.37
PubChem CID 19081435
Formula Weight 316.38
Percent Purity 97%
Chemical Name or Material 2-(1-Benzofuran-3-yl)ethyl 4-methylbenzenesulfonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.