missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Hydroxyethyl) Promazine Sulfoxide (mixture of diastereomers), TRC

Product Code. 30133382
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133382 100 mg 100mg
30006504 10 mg 10mg
30010670 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133382 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCH942290100MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C19H24N2O2S
Molecular Weight (g/mol) 344.47
Synonym 2-(1-Hydroxyethyl) Promazine Sulfoxide
IUPAC Name 1-[10-[3-(dimethylamino)propyl]-5-oxophenothiazin-2-yl]ethanol
SMILES CC(O)c1ccc2c(c1)N(CCCN(C)C)c3ccccc3S2=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H24N2O2S/c1-14(22)15-9-10-19-17(13-15)21(12-6-11-20(2)3)16-7-4-5-8-18(16)24(19)23/h4-5,7-10,13-14,22H,6,11-12H2,1-3H3
Formula Weight 344.1558
Percent Purity >95%
Chemical Name or Material 2-(1-Hydroxyethyl) Promazine Sulfoxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.