missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Methyl-2-phenyl-ethylimin)-5-hydroxymethyl-furfural, TRC

Product Code. 30477731
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477731 250 mg 250mg
30466260 50 mg 50mg
30465502 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477731 Supplier Toronto Research Chemicals Supplier No. TRCM558420250MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53847-95-3
Molecular Formula C15H17NO2
Molecular Weight (g/mol) 243.3
Synonym (5-(((1-phenylpropan-2-yl)imino)methyl)furan-2-yl)methanol
IUPAC Name [5-[(Z)-1-phenylpropan-2-yliminomethyl]furan-2-yl]methanol
SMILES CC(Cc1ccccc1)\N=C/c2oc(CO)cc2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H17NO2/c1-12(9-13-5-3-2-4-6-13)16-10-14-7-8-15(11-17)18-14/h2-8,10,12,17H,9,11H2,1H3/b16-10-
Formula Weight 243.1259
Chemical Name or Material 2-(1-Methyl-2-phenyl-ethylimin)-5-hydroxymethyl-furfural
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.