missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Naphthyl)ethanoyl chloride, 97%, Thermo Scientific™

Product Code. 10591883
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10591883 250 mg 250mg
10724773 1 g 1g
10265594 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10591883 Supplier Thermo Scientific Maybridge Supplier No. S07397CB
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 5121-00-6
Molecular Formula C12H9ClO
Molecular Weight (g/mol) 204.65
MDL Number MFCD00175852
InChI Key DSVAZLXLRDXHKO-UHFFFAOYSA-N
Synonym 2-1-naphthyl ethanoyl chloride, 2-naphthalen-1-yl acetyl chloride, 1-naphthylacetyl chloride, naphth-1-ylacetyl chloride, 1-naphthaleneacetyl chloride, naphth-1-yl acetyl chloride, napthylacetyl chloride, acmc-20aork, 1-naphthylacetylchloride, 1-naphthaleneacetylchloride
PubChem CID 2795386
IUPAC Name 2-naphthalen-1-ylacetyl chloride
SMILES ClC(=O)CC1=CC=CC2=CC=CC=C12

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 204.66
Percent Purity 95%
Chemical Name or Material 2-(1-Naphthyl)ethanoyl chloride
compliance-icons
Product Identifier
  • 2-(1-Naphthyl)ethanoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.