missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Piperazinyl)pyrimidine, 98+%

Product Code. 10011310 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10011310 1 g Glass bottle 1g
10337210 5 g Glass bottle 5g
10519633 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10011310 Supplier Thermo Scientific Acros Supplier No. 134320010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 20980-22-7 | C8H12N4 | 164.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20980-22-7
Molecular Formula C8H12N4
Molecular Weight (g/mol) 164.21
MDL Number MFCD00040742
InChI Key MRBFGEHILMYPTF-UHFFFAOYSA-N
Synonym 2-1-piperazinyl pyrimidine, 1-2-pyrimidyl piperazine, 2-piperazin-1-yl pyrimidine, 1-2-pyrimidinyl piperazine, 2-piperazinopyrimidine, campiron, pyrimidine, 2-1-piperazinyl, campirone, kampirone, 2-piperazin-1-yl-pyrimidine
PubChem CID 88747
IUPAC Name 2-piperazin-1-ylpyrimidine
SMILES C1CN(CCN1)C2=NC=CC=N2

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 29°C to 36°C
Color Yellow
Density 1.0700g/mL
Boiling Point 96°C to 97°C (0.2 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5850 to 1.588
Quantity 1 g
Beilstein 25, IV, 2083
Specific Gravity 1.07
Formula Weight 164.21
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material 2-(1-Piperazinyl)pyrimidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.