missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Piperazinyl)pyrimidine, 99%

Product Code. 11342807
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342807 100 g 100g
11322807 5 g 5g
11332807 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11342807 Supplier Thermo Scientific Alfa Aesar Supplier No. L15884.22
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20980-22-7 | C8H12N4 | 164.212 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20980-22-7
Molecular Formula C8H12N4
Molecular Weight (g/mol) 164.212
MDL Number MFCD00040742
InChI Key MRBFGEHILMYPTF-UHFFFAOYSA-N
Synonym 2-1-piperazinyl pyrimidine, 1-2-pyrimidyl piperazine, 2-piperazin-1-yl pyrimidine, 1-2-pyrimidinyl piperazine, 2-piperazinopyrimidine, campiron, pyrimidine, 2-1-piperazinyl, campirone, kampirone, 2-piperazin-1-yl-pyrimidine
PubChem CID 88747
IUPAC Name 2-piperazin-1-ylpyrimidine
SMILES C1CN(CCN1)C2=NC=CC=N2

Specifications

Melting Point 31°C to 35°C
Boiling Point 118°C to 120°C (2 mmHg)
Quantity 100 g
UN Number UN3259
Beilstein 151178
Sensitivity Air sensitive; Hygroscopic
Formula Weight 164.21
Percent Purity 99%
Chemical Name or Material 2-(1-Piperazinyl)pyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.