missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Pyrrolidinyl)nicotinaldehyde, 97%, Thermo Scientific™

Product Code. 10300883
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10300883 1 g Amber Glass Bottle 1g
10269052 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10300883 Supplier Thermo Scientific Maybridge Supplier No. MO07059DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-39-4
Molecular Formula C10H12N2O
Molecular Weight (g/mol) 176.22
MDL Number MFCD05865136
InChI Key FTACFSVJFQMXQE-UHFFFAOYSA-N
Synonym 2-1-pyrrolidinyl nicotinaldehyde, 2-pyrrolidin-1-yl nicotinaldehyde, 3-pyridinecarboxaldehyde,2-1-pyrrolidinyl, 2-pyrrolidin-1-yl pyridine-3-carbaldehyde, 2-pyrrolidinylpyridine-3-carbaldehyde, 2-pyrrolidin-1-yl-pyridine-3-carbaldehyde, 2-1-pyrrolidinyl-3-pyridinecarboxaldehyde, 3-pyridinecarboxaldehyde, 2-1-pyrrolidinyl
PubChem CID 2794763
IUPAC Name 2-pyrrolidin-1-ylpyridine-3-carbaldehyde
SMILES O=CC1=C(N=CC=C1)N1CCCC1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 176.22
Percent Purity 97%
Chemical Name or Material 2-(1-Pyrrolidinyl)nicotinaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.