missing translation for 'onlineSavingsMsg'
Learn More

2-[1-(tert-Butoxycarbonyl)piperid-4-yl]-1,3-thiazole-4-carboxylic acid, ≥95%, Thermo Scientific™

Product Code. 10768643
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10768643 250 mg Amber Glass Bottle 250mg
10471893 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10768643 Supplier Thermo Scientific Maybridge Supplier No. CC31001CB
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 668484-45-5
Molecular Formula C13H19N3O4S
Molecular Weight (g/mol) 313.372
MDL Number MFCD06658980
InChI Key FMSFMUBALWPCQM-UHFFFAOYSA-N
Synonym 2-4-tert-butoxycarbonyl piperazin-1-yl thiazole-4-carboxylic acid, 2-4-tert-butoxycarbonyl piperazin-1-yl-1,3-thiazole-4-carboxylic acid
PubChem CID 59496772
IUPAC Name 2-[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]-1,3-thiazole-4-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=NC(=CS2)C(=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 312.39
Percent Purity 95%
Chemical Name or Material 2-[1-(Tert-butoxycarbonyl)-4-piperidinyl]-1, 3-thiazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.