missing translation for 'onlineSavingsMsg'
Learn More

2-[(11bR)-3H-Binaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylamine, 97+%, Thermo Scientific Chemicals

Product Code. 15425746
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425746 250 mg 250mg
15415746 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425746 Supplier Thermo Scientific Alfa Aesar Supplier No. H60423.MD
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1092064-00-0 | Molecular Formula: C24H22NP | Molecular Weight: 355.421 g/mol

2-[(11bR)-3H-Binaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylamine, is used as a application of innovative catalyst, ligand synthesis and application in asymmetric catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-[(11bR)-3H-Binaphtho[2,1-c:1′,2′-e]phosphepin-4(5H)-yl]ethylamine, is used as a application of innovative catalyst, ligand synthesis and application in asymmetric catalysis.

Solubility
Soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Air, sensitive.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1092064-00-0
Molecular Formula C24H22NP
Molecular Weight (g/mol) 355.421
MDL Number MFCD11045442
InChI Key PXDLCEFIOZLJLW-UHFFFAOYSA-N
Synonym 2-11br-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethyl amine, 2-11br-3h-binaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylamine, 2-11bs-3h-binaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethanamine, 2-11bs-3h-binaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethanamine, kanata purity, 2-11bs-3h-binaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylamine, 2-11bs-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethyl amine, 2-11bs-3h-binaphtho 2,1-c:1 inverted exclamation marka,2 inverted exclamation marka-e phosphepin-4 5h-yl ethanamine, 2-13-phosphapentacyclo 13.8.0.0 2 , 1 1 .0 3 ,?.0 1 ?, 2 3 tricosa-1 15 ,2 11 ,3,5,7,9,16,18,20,22-decaen-13-yl ethanamine, 2-13-phosphapentacyclo 13.8.0.0_,__.0_,?.0_?,__ tricosa-1 15 ,2,4,6,8,10,16,18,20,22-decaen-13-yl ethanamine
PubChem CID 46177751
SMILES C1C2=C(C3=CC=CC=C3C=C2)C4=C(CP1CCN)C=CC5=CC=CC=C54

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 355.41
Percent Purity ≥97%
Chemical Name or Material 2-[(11bR)-3H-Binaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.