missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-1,2,4-triazol-1-ylmethyl)aniline, 97%, Thermo Scientific™

Product Code. 10205764
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10205764 1 g 1g
10133474 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10205764 Supplier Thermo Scientific Maybridge Supplier No. CC47514DA
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 127988-21-0
Molecular Formula C9H10N4
Molecular Weight (g/mol) 174.207
MDL Number MFCD09025845
InChI Key CBGQHZNXHUCAGE-UHFFFAOYSA-N
Synonym 2-1h-1,2,4-triazol-1-ylmethyl aniline, 2-1,2,4-triazol-1-yl methyl aniline, 2-1,2,4-triazol-1-ylmethyl aniline, 2-1h-1,2,4-triazol-1-ylmethyl benzeneamine, benzenamine,2-1h-1,2,4-triazol-1-ylmethyl, acmc-20msof, 2-1,2,4-triazolylmethyl phenylamine
PubChem CID 6482003
IUPAC Name 2-(1,2,4-triazol-1-ylmethyl)aniline
SMILES C1=CC=C(C(=C1)CN2C=NC=N2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 174.21
Percent Purity 97%
Chemical Name or Material 2-(1H-1, 2, 4-Triazol-1-ylmethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.