missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-1,2,4-triazol-1-ylmethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10604015
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10604015 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10604015 Supplier Thermo Scientific Maybridge Supplier No. CC47501DA
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 876718-01-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H9N3O2
MDL Number MFCD07186455
Quantity 1 g
Synonym 2-1h-1,2,4-triazol-1-ylmethyl benzoic acid, 2-1,2,4 triazol-1-ylmethyl-benzoic acid, 2-1,2,4-triazol-1-ylmethyl benzoic acid, 2-1,2,4-triazolylmethyl benzoic acid, 2-1h-1,2,4-triazol-1-yl methyl benzoic acid
InChI Key KPKNLIVZZRDKGI-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=CC=C1CN1C=NC=N1
IUPAC Name 2-[(1H-1,2,4-triazol-1-yl)methyl]benzoic acid
Molecular Weight (g/mol) 203.20
PubChem CID 3159712
Formula Weight 203.2
Percent Purity 97%
Chemical Name or Material 2-(1H-1, 2, 4-Triazol-1-ylmethyl)benzoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.