missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-Imidazol-1-yl)acetonitrile, 97%, BLD Pharm™

Product Code. 31340215
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
500g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31340215 1 g -
31339457 500 g 500g
31339687 10 g -
31338464 5 g -
31337761 100 g -
31340181 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31340215 Supplier BLD Pharm Supplier No. BD1344511g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 98873-55-3
Molecular Formula C5H5N3
MDL Number MFCD06421433
Quantity 1 g
SMILES N#CCN1C=CN=C1
Molecular Weight (g/mol) 107.11
Percent Purity 97%
Chemical Name or Material 2-(1H-Imidazol-1-yl)acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.