missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-Pyrazol-1-yl)benzoic acid, ≥97%, Thermo Scientific™

Product Code. 10719773
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10719773 1 g 1g
10435032 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10719773 Supplier Thermo Scientific Maybridge Supplier No. CC24401DA
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 55317-53-8
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD03086184
InChI Key MHACZVWKWUMHRR-UHFFFAOYSA-N
Synonym 2-1h-pyrazol-1-yl benzoic acid, 2-1-pyrazolyl benzoic acid, 2-pyrazol-1-yl benzoic acid, benzoic acid, 2-1h-pyrazol-1-yl, 2-pyrazol-1-yl-benzoic acid, 2-pyrazolylbenzoic acid, pubchem19561, 2-1-pyrazolyl benzoicacid, benzoic acid,2-1h-pyrazol-1-yl
PubChem CID 4738383
IUPAC Name 2-pyrazol-1-ylbenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)N2C=CC=N2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material 2-(1H-pyrazol-1-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.