missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-Pyrazol-3-yl)pyridine, 98%

Product Code. 11450210
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450210 1 g 1g
11460210 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450210 Supplier Thermo Scientific Alfa Aesar Supplier No. H50670.03
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 75415-03-1 | C8H7N3 | 145.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75415-03-1
Molecular Formula C8H7N3
Molecular Weight (g/mol) 145.17
MDL Number MFCD00115151
InChI Key HKEWOTUTAYJWQJ-UHFFFAOYSA-N
Synonym 2-1h-pyrazol-3-yl pyridine, 2-1h-pyrazol-5-yl pyridine, 2-pyrazol-3-yl-pyridine, 2-1h-pyrazol-3-yl-pyridine, 2-3-pyrazolyl pyridine, pyridine, 2-1h-pyrazol-3-yl, 3-2-pyridyl pyrazole, 2-2h-pyrazol-3-yl pyridine, 2-3e-pyrazol-3-ylidene-1h-pyridine, pyrazolylpyridine
PubChem CID 2797657
IUPAC Name 2-(1H-pyrazol-5-yl)pyridine
SMILES N1N=CC=C1C1=CC=CC=N1

Specifications

Melting Point 121°C to 122°C
Quantity 1 g
Formula Weight 145.16
Percent Purity 98%
Chemical Name or Material 2-(1H-Pyrazol-3-yl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.