missing translation for 'onlineSavingsMsg'
Learn More

2-[2-[2-[2-(4-Dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethoxy]ethoxy]ethanol, TRC

Product Code. 30134663
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134663 250 mg 250mg
30004960 100 mg 100mg
30008997 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134663 Supplier Toronto Research Chemicals Supplier No. TRCD418685250MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1356906-17-6
Molecular Formula C25 H33 N3 O4 S
Molecular Weight (g/mol) 471.612
Synonym 2-[2-[2-[2-(4-Dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethoxy]ethoxy]ethanol, Ethanol, 2-[2-[2-[2-(4-dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethoxy]ethoxy]-, Quetiapine Fumarate Imp. J (EP)
IUPAC Name 2-[2-[2-[2-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)ethoxy]ethoxy]ethoxy]ethanol
SMILES OCCOCCOCCOCCN1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C25H33N3O4S/c29-14-16-31-18-20-32-19-17-30-15-13-27-9-11-28(12-10-27)25-21-5-1-3-7-23(21)33-24-8-4-2-6-22(24)26-25/h1-8,29H,9-20H2
Formula Weight 471.219
Chemical Name or Material 2-[2-[2-[2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethoxy]ethoxy]ethoxy]ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.