missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(2,2,2-Trimethylacetamido)nicotinic acid

Product Code. 11424370
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424370 5 g 5g
11414370 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424370 Supplier Alfa Aesar™ Supplier No. H50008.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 125867-25-6
Molecular Formula C11H14N2O3
Molecular Weight (g/mol) 222.244
MDL Number MFCD03659710
InChI Key KXLOGMIKCNUSNG-UHFFFAOYSA-N
Synonym 2-pivalamidonicotinic acid, 2-2,2-dimethyl-propionylamino-nicotinic acid, 2-2,2,2-trimethylacetamido nicotinic acid, 2-2,2-dimethylpropanoyl amino nicotinic acid, 2-2,2-dimethylpropanamido pyridine-3-carboxylic acid, 2-pivaloylamino nicotinic acid, 2-2,2-dimethylpropanoylamino pyridine-3-carboxylic acid, maybridge3_004279, acmc-20aorg, pubchem16551
PubChem CID 2779758
IUPAC Name 2-(2,2-dimethylpropanoylamino)pyridine-3-carboxylic acid
SMILES CC(C)(C)C(=O)NC1=C(C=CC=N1)C(=O)O

Specifications

Quantity 5 g
Formula Weight 222.25
Chemical Name or Material 2-(2,2,2-Trimethylacetamido)nicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.