missing translation for 'onlineSavingsMsg'
Learn More

2-(2-(2-(4-((4-Chlorophenyl)(phenyl)methyl)piperazin-1-yl)ethoxy)ethoxy)acetic Acid tert-Butyl Ester, TRC

Product Code. 30349286
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30349286 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30349286 Supplier Toronto Research Chemicals Supplier No. TRCC375045100MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2-(2-(4-((4-Chlorophenyl)(phenyl)methyl)piperazin-1-yl)ethoxy)ethoxy)acetic Acid tert-Butyl Ester
CAS 2518202-76-9
Molecular Formula C27 H37 Cl N2 O4
Quantity 100 mg
InChI Formula InChI=1S/C27H37ClN2O4/c1-27(2,3)34-25(31)21-33-20-19-32-18-17-29-13-15-30(16-14-29)26(22-7-5-4-6-8-22)23-9-11-24(28)12-10-23/h4-12,26H,13-21H2,1-3H3
SMILES CC(C)(C)OC(=O)COCCOCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3
IUPAC Name tert-butyl 2-[2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]ethoxy]acetate
Molecular Weight (g/mol) 489.047
Formula Weight 488.244
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.