missing translation for 'onlineSavingsMsg'
Learn More

2-(2-((2-(((4-Chlorophenyl)(phenyl)methyl)amino)ethyl)amino)ethoxy)ethanol, TRC

Product Code. 30354040
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30354040 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354040 Supplier Toronto Research Chemicals Supplier No. TRCC29295025MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2-((2-(((4-Chlorophenyl)(phenyl)methyl)amino)ethyl)amino)ethoxy)ethanol
CAS 2518402-06-5
Molecular Formula C19 H25 Cl N2 O2
Quantity 25 mg
InChI Formula InChI=1S/C19H25ClN2O2/c20-18-8-6-17(7-9-18)19(16-4-2-1-3-5-16)22-11-10-21-12-14-24-15-13-23/h1-9,19,21-23H,10-15H2
SMILES OCCOCCNCCNC(c1ccccc1)c2ccc(Cl)cc2
IUPAC Name 2-[2-[2-[[(4-chlorophenyl)-phenylmethyl]amino]ethylamino]ethoxy]ethanol
Molecular Weight (g/mol) 348.867
Formula Weight 348.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.