missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(2-chloroethoxy)ethyl]-1H-isoindole-1,3(2H)-dione, 97%, Maybridge

Product Code. 10374192
Change view
Click to view available options
:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10374192 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10374192 Supplier Thermo Scientific Maybridge Supplier No. KM10098DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 58290-51-0
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C12H12ClNO3
MDL Number MFCD00196075
Quantity 1 g
Synonym 2-2-2-chloroethoxy ethyl-1h-isoindole-1,3 2h-dione, 2-2-2-chloroethoxy ethyl isoindole-1,3-dione, 2-2-2-chloroethoxy ethyl-2,3-dihydro-1h-isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2-2-chloroethoxy ethyl, n-2-2-chloroethoxy ethyl phthalimide, 2-2-2-chloroethoxy ethyl isoindoline-1,3-dione, 2-2-2-chloroethoxy ethyl-1h-isoindole-1,3 2h dione, 2-2-2-chloroethoxy ethyl benzo c azolidine-1,3-dione
InChI Key DCWWMFNXTOGDNJ-UHFFFAOYSA-N
SMILES ClCCOCCN1C(=O)C2=CC=CC=C2C1=O
IUPAC Name 2-[2-(2-chloroethoxy)ethyl]isoindole-1,3-dione
Molecular Weight (g/mol) 253.68
PubChem CID 2735735
Formula Weight 253.68
Percent Purity 97%
Chemical Name or Material 2-[2-(2-Chloroethoxy)ethyl]-1H-isoindole-1, 3(2H)-dione
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.