missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trichloroethanol, 99%

Product Code. 11348015
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348015 100 g 100g
11338015 5 g 5g
15468879 25 g 25g
11358015 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11348015 Supplier Thermo Scientific Alfa Aesar Supplier No. L08163.22
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 115-20-8 | C2H3Cl3O | 149.395 g/mol

2,2,2-Trichloroethanol is used as a sedative or a hypnotic. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trichloroethanol is used as a sedative or a hypnotic. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with oxidizing agents, Strong acids, Strong bases, Strong reducing agents, Acid chlorides, Acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-20-8
Molecular Formula C2H3Cl3O
Molecular Weight (g/mol) 149.395
MDL Number MFCD00004677
InChI Key KPWDGTGXUYRARH-UHFFFAOYSA-N
Synonym trichloroethanol, trichlorethanol, trichloroethyl alcohol, ethanol, 2,2,2-trichloro, 2,2,2-trichloro-1-ethanol, hydroxymethyl trichloromethane, 2,2,2-trichloroethyl alcohol, 2,2,2-trichloroethan-1-ol, unii-aw835aj62n, beta-trichloroethanol
PubChem CID 8259
ChEBI CHEBI:28094
IUPAC Name 2,2,2-trichloroethanol
SMILES C(C(Cl)(Cl)Cl)O

Specifications

Melting Point 17°C to 18°C
Density 1.56
pH 5 to 6
Boiling Point 151°C to 153°C
Flash Point >110°C (230°F)
Refractive Index 1.4895
Quantity 100 g
Beilstein 1697495
Sensitivity Hygroscopic
Merck Index 14,9638
Solubility Information Slightly soluble in water.
Formula Weight 149.4
Percent Purity 99%
Chemical Name or Material 2,2,2-Trichloroethanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,2,2-Trichloroethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2,2,2-Trichloroethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.