missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15448175
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15448175 1 g 1g
15458175 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 15448175

Brand: Thermo Scientific Alfa Aesar H32717.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1138011-22-9
Molecular Formula C7H8ClF3N2
Molecular Weight (g/mol) 212.6
MDL Number MFCD06739079
InChI Key NLDVAGWIUPSGQC-UHFFFAOYSA-N
Synonym 2,2,2-trifluoro-1-pyridin-3-yl ethanamine hydrochloride, +/--2,2,2-trifluoro-1-pyridin-3-ylethylamine hydrochloride, 2,2,2-trifluoro-1-pyridin-3-ylethylamine hydrochloride, 2,2,2-trifluoro-1-pyridin-3-yl-ethylamine hydrochloride, 2,2,2-trifluoro-1-3-pyridyl ethylamine hydrochloride, acmc-20aokz, 2,2,2-trifluoro-1-3-pyridinyl ethanamine hydrochloride, 2,2,2-trifluoro-1-3-pyridinyl ethylamine hydrochloride, 2,2,2-tris fluoranyl-1-pyridin-3-yl-ethanamine hydrochloride
PubChem CID 45594310
IUPAC Name 2,2,2-trifluoro-1-pyridin-3-ylethanamine;hydrochloride
SMILES C1=CC(=CN=C1)C(C(F)(F)F)N.Cl

Specifications

Melting Point 97°C to 102°C
Quantity 1 g
Formula Weight 212.6
Percent Purity 95%
Chemical Name or Material 2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.