missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethanol, ≥99.8% (GC), Eluent additive for LC-MS, Solstice

Product Code. 15670800 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 mL
10 x 1 mL
Packaging:
Ampoule
Glass Bottle
Unit Size:
50mL
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15670800 50 mL Glass Bottle 50mL
15628500 10 x 1 mL Ampoule Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15670800 Supplier Honeywell-Fluka Supplier No. 0584150ML
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eluent additive for LC-MS

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-89-8
Molecular Formula C2H3F3O
Molecular Weight (g/mol) 100.04
MDL Number MFCD00004672
InChI Key RHQDFWAXVIIEBN-UHFFFAOYSA-N
Synonym trifluoroethanol, ethanol, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-ol, fluorinol 85, 2,2,2-trifluoroethyl alcohol, trifluoroethyl alcohol, trifluoro ethanol, tfe, perfluoro-1,1-dihydroethanol, tfetoh
PubChem CID 6409
ChEBI CHEBI:42330
IUPAC Name 2,2,2-trifluoroethanol
SMILES OCC(F)(F)F

Specifications

Melting Point -44°C (literature)
Color Colorless
Density 1.391g/mL at 20°C, 1.373g/mL at 25°C
Boiling Point 77°C to 80°C
Flash Point 29°C (84.2°F)
Packaging Glass Bottle
Linear Formula CF3CH2OH
Refractive Index n20/D 1.3(lit.)
Quantity 50 mL
UN Number UN1986
Beilstein 1733203
Vapor Density 3.5 (vs air)
Formula Weight 100.04g/mol
Vapor Pressure 70mmHg (25°C)
Chemical Name or Material 2,2,2-Trifluoroethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.