missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethanol, ≥99%, Reagent Grade, Honeywell™

Product Code. 15638190 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15638190 500 g Glass bottle 500g
15628190 25 g Glass bottle 25g
15618190 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15638190 Supplier Honeywell Research Chemicals Supplier No. T63002500G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-89-8
Molecular Formula C2H3F3O
Molecular Weight (g/mol) 100.04
MDL Number MFCD00004672
InChI Key RHQDFWAXVIIEBN-UHFFFAOYSA-N
Synonym trifluoroethanol, ethanol, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-ol, fluorinol 85, 2,2,2-trifluoroethyl alcohol, trifluoroethyl alcohol, trifluoro ethanol, tfe, perfluoro-1,1-dihydroethanol, tfetoh
PubChem CID 6409
ChEBI CHEBI:42330
SMILES OCC(F)(F)F

Specifications

Melting Point -44°C
Color Colorless
Density 1.373g/mL at 25°C
Boiling Point 73°C to 77°C
Flash Point 29°C (84.2°F)
Odor Characteristic
Packaging Glass bottle
Linear Formula CF3CH2OH
Quantity 500 g
UN Number UN1986
Beilstein 1733203
IUPAC Name 2,2,2-trifluoroethanol
Vapor Density 3.5 (vs air)
Vapor Pressure 70mmHg (25°C)
Percent Purity ≥99%
Chemical Name or Material 2,2,2-Trifluoroethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.