missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals

Product Code. 10093680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10093680 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10093680 Supplier Thermo Scientific Acros Supplier No. 312780250
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352-87-4
Molecular Formula C6H7F3O2
Molecular Weight (g/mol) 168.12
MDL Number MFCD00013576,MFCD00274739
InChI Key QTKPMCIBUROOGY-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl methacrylate, trifluoroethyl methacrylate, methacrylic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethylmethacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoroethyl ester, 2-methyl-acrylic acid 2,2,2-trifluoro-ethyl ester, 2-propenoic acid, 2-methyl-, trifluoroethyl ester, 2-methyl-2-propenoic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl m, acmc-1af8z
PubChem CID 9608
IUPAC Name 2,2,2-trifluoroethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(F)(F)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Density 1.1800g/mL
Boiling Point 59.0°C (100.0 mmHg)
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CO2CH2CF3
Refractive Index 1.3600 to 1.3620
Quantity 25 g
Specific Gravity 1.18
Stabilizer 100ppm MEHQ
Formula Weight 168.12
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2,2,2-Trifluoroethyl methacrylate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.