missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trimethylthioacetamide, 97%

Product Code. 11370036
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11370036 1 g 1g
11360036 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11370036 Supplier Thermo Scientific Alfa Aesar Supplier No. H32092.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 630-22-8 | C5H11NS | 117.21 g/mol

2,2,2-Trimethylthioacetamide, is used as a pharmaceutical and organic intermediate, an important amino protective agent, pharmaceutical and synthetic organic make intermediates, organic synthesis used to introduce t- Boc protect gene species.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trimethylthioacetamide, is used as a pharmaceutical and organic intermediate, an important amino protective agent, pharmaceutical and synthetic organic make intermediates, organic synthesis used to introduce t- Boc protect gene species.

Solubility
Soluble in water (10 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-22-8
Molecular Formula C5H11NS
Molecular Weight (g/mol) 117.21
MDL Number MFCD09742834
InChI Key FJZJUSOFGBXHCV-UHFFFAOYSA-N
Synonym 2,2,2-trimethylthioacetamide, 2,2-dimethylthiopropionamide, neopentanethioamide, propanethioamide, 2,2-dimethyl, thiopivalamide, 2,2-dimethyl-thiopropionamide, 2,2-dimethylthio propionamide, propanethioamide,2,2-dimethyl, 2-methyl-2-methylthiopropionamide, propionamide, 2-methyl-2-methylthio
PubChem CID 3031130
IUPAC Name 2,2-dimethylpropanethioamide
SMILES CC(C)(C)C(=S)N

Specifications

Melting Point 113°C to 117°C
Quantity 1 g
Solubility Information Soluble in water (10g/L) (25°C).
Formula Weight 117.21
Percent Purity 97%
Chemical Name or Material 2,2,2-Trimethylthioacetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.