missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,3-Pentafluoro-1-propanol, 97%

Product Code. p-178665 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10790581

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 422-05-9 | C3H3F5O | 150.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 422-05-9
Molecular Formula C3H3F5O
Molecular Weight (g/mol) 150.05
MDL Number MFCD00004673
InChI Key PSQZJKGXDGNDFP-UHFFFAOYSA-N
Synonym 2,2,3,3,3-pentafluoro-1-propanol, pentafluoro-1-propanol, 1,1-dihydroperfluoropropanol, pfpoh, 2,2,3,3,3-pentafluoropropanol, 1h,1h-pentafluoropropanol, 1-propanol, 2,2,3,3,3-pentafluoro, 1h,1h-pentafluoropropanol-1, c2f5ch2oh, unii-epj1s961sn
PubChem CID 9872
IUPAC Name 2,2,3,3,3-pentafluoropropan-1-ol
SMILES C(C(C(F)(F)F)(F)F)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White to Yellow
Boiling Point 81.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CF3CF2CH2OH
Refractive Index 1.2880 to 1.2950
Quantity 5 g
Beilstein 01, IV, 1438
Formula Weight 150.05
Percent Purity 97%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 2,2,3,3,3-Pentafluoro-1-propanol, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.