missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,3-Pentafluoropropionamide, 97%, Thermo Scientific™

Product Code. 11447164
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447164 25 g 25g
11437164 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11447164 Supplier Thermo Scientific Alfa Aesar Supplier No. B21566.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,2,3,3,3-Pentafluoropropionamide, is used as a chemical, organic, pharamceutical intermediate.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 354-76-7
Molecular Formula C3H2F5NO
Molecular Weight (g/mol) 163.05
MDL Number MFCD00039771
InChI Key KQTOYEUYHXUEDB-UHFFFAOYSA-N
PubChem CID 67722
IUPAC Name pentafluoropropanamide
SMILES NC(=O)C(F)(F)C(F)(F)F

Specifications

Melting Point 92°C to 97°C
Quantity 25 g
Beilstein 1773386
Synonym pentafluoropropionamide, pentafluoropropanamide, propanamide, 2,2,3,3,3-pentafluoro, 2,2,3,3,3-pentafluoropropionamide, acmc-1cjql, pentafluoro propionic acid amide, 2,2,3,3,3-pentafluoropropionamide,, propanamide,2,2,3,3,3-pentafluoro
Solubility Information Soluble in methanol.
Formula Weight 163.05
Percent Purity 97%
Chemical Name or Material 2,2,3,3,3-Pentafluoropropionamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.