missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,4-Heptafluorobutylamine, 97%

Product Code. 10222912 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10222912 1 g Glass bottle 1g
10792833 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10222912 Supplier Thermo Scientific Acros Supplier No. 368770010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. Rückgaberichtlinie anzeigen

CAS: 374-99-2 | C4H4F7N | 199.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 374-99-2
Molecular Formula C4H4F7N
Molecular Weight (g/mol) 199.07
MDL Number MFCD00014817
InChI Key WBGBQSRNXPVFDB-UHFFFAOYSA-N
PubChem CID 67807
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutan-1-amine
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)N

Spezifikation

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4930g/mL
Boiling Point 70°C to 71°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CF3CF2CF2CH2NH2
Refractive Index 1.2960 to 1.298
Quantity 1 g
Specific Gravity 1.493
Synonym 2,2,3,3,4,4,4-heptafluorobutylamine, 1h,1h-heptafluorobutylamine, heptafluorobutylamine, butylamine, 2,2,3,3,4,4,4-heptafluoro, 1-butanamine, 2,2,3,3,4,4,4-heptafluoro, 1h,1h-perfluorobutylamine, acmc-1af94, 1-butanamine,2,2,3,3,4,4,4-heptafluoro
Formula Weight 199.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluorobutylamine, 97%
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 2,2,3,3,4,4,4-Heptafluorobutylamine
Signalwort
  • Danger
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Gefahrenhinweis
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.