missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate, tech.

Product Code. 11334344
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334344 5 g 5g
11324344 1 g 1g
11344344 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334344 Supplier Thermo Scientific Alfa Aesar Supplier No. L00875.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 375-02-0 | C4HF7O | 198.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-02-0
Molecular Formula C4HF7O
Molecular Weight (g/mol) 198.04
MDL Number MFCD00039731
InChI Key IQJZGNJYXIIMGP-UHFFFAOYSA-N
Synonym heptafluorobutyraldehyde, butanal, heptafluoro, butyraldehyde, heptafluoro, heptafluorobutyraldehyde, tech., perfluorobutanal, perfluorobutyraldehyde, c3f7cho, heptafluoro-n-butyraldehyde, butanal,2,2,3,3,4,4,4-heptafluoro
PubChem CID 67809
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanal
SMILES FC(F)(F)C(F)(F)C(F)(F)C=O

Specifications

Refractive Index 1.32
Quantity 5 g
Beilstein 1367365
Formula Weight 198.04 (Anhydrous)
Grade Technical
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate
compliance-icons
Product Identifier
  • 2,2,3,3,4,4,4-Heptafluorobutyraldehydehydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Heptafluorobutyraldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.