missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol, 97%

Product Code. 11399225
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399225 5 g 5g
11309235 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399225 Supplier Thermo Scientific Alfa Aesar Supplier No. L16560.06
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 355-74-8 | C6H6F8O2 | 262.099 g/mol

2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 355-74-8
Molecular Formula C6H6F8O2
Molecular Weight (g/mol) 262.099
MDL Number MFCD00069087
InChI Key NHEKBXPLFJSSBZ-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,5,5-octafluoro-1,6-hexanediol, 1,6-dihydroxy-2,2,3,3,4,4,5,5-octafluorohexane, 1,6-hexanediol, 2,2,3,3,4,4,5,5-octafluoro, 2,2,3,3,4,4,5,5-octafluorohexan-1,6-diol, acmc-1code, nhekbxplfjssbz-uhfffaoysa, 2,2,3,3,4,4,5,5octafluoro-1,6-hexanediol
PubChem CID 136181
IUPAC Name 2,2,3,3,4,4,5,5-octafluorohexane-1,6-diol
SMILES C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 67°C to 70°C
Boiling Point 100°C (3 mmHg)
Quantity 5 g
Beilstein 1784039
Solubility Information Soluble in water.
Formula Weight 262.1
Percent Purity 97%
Chemical Name or Material 2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.