missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,5,5-Octafluoro-1-pentanol, 98%

Product Code. 11470621
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470621 100 g 100g
11460621 25 g 25g
11480621 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470621 Supplier Thermo Scientific Alfa Aesar Supplier No. B20108.22
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 355-80-6 | C5H4F8O | 232.07 g/mol

2,2,3,3,4,4,5,5-Octafluoro-1-pentanol is used as a cosurfactant in the synthesis of silver and silver iodide nanocrystals. It was also used in the synthesis of Ag2S nanocrystals with a characteristic surface plasmon resonance absorption at 330nm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol is used as a cosurfactant in the synthesis of silver and silver iodide nanocrystals. It was also used in the synthesis of Ag2S nanocrystals with a characteristic surface plasmon resonance absorption at 330nm.

Solubility
Insoluble in water.

Notes
Store away from air. Store under dry inert gas. Incompatible with oxidizing agents and air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 355-80-6
Molecular Formula C5H4F8O
Molecular Weight (g/mol) 232.07
MDL Number MFCD00039631
InChI Key JUGSKHLZINSXPQ-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,5,5-octafluoro-1-pentanol, 1h,1h,5h-octafluoro-1-pentanol, 1-pentanol, 2,2,3,3,4,4,5,5-octafluoro, 1,1,5-trihydrooctafluoropentyl alcohol, 2,2,3,3,4,4,5,5-octafluoropentanol, 1,1,5-trihydrooctafluoropentan-1-ol, unii-d05k8ejx15, 1h,1h,5h-octafluoropentanol, 2,2,3,3,4,4,5,5-octafluoropentyl alcohol, alpha,alpha,omega-trihydroperfluoropentanol
PubChem CID 9641
IUPAC Name 2,2,3,3,4,4,5,5-octafluoropentan-1-ol
SMILES OCC(F)(F)C(F)(F)C(F)(F)C(F)F

Specifications

Melting Point <-50°C
Density 1.667
Boiling Point 141°C to 142°C
Flash Point 75°C (167°F)
Refractive Index 1.318
Quantity 100 g
Beilstein 1773494
Merck Index 14,6746
Solubility Information Insoluble in water.
Formula Weight 232.07
Percent Purity 98%
Chemical Name or Material 2,2,3,3,4,4,5,5-Octafluoro-1-pentanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.