missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt, 98+ atom % D, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt is used as a deuterium labeled silane. Also used in pharmaceutical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt is used as a deuterium labeled silane. Also used in pharmaceutical industry.
Solubility
Soluble in water.
Notes
It is hygroscopic. Incompatible with oxidizing agents.
2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt is used as a deuterium labeled silane. Also used in pharmaceutical industry.
Solubility
Soluble in water.
Notes
It is hygroscopic. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | >300°C |
Linear Formula | (CH3)3SiCD2CD2CO2Na |
Quantity | 1 g |
Beilstein | 4046239 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 172.27 |
Assay | ≥98 atom %D |
Chemical Name or Material | 2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'