missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3-Tetrafluoro-1-propanol, 97%

Product Code. 11477204
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477204 10 g 10g
11487204 50 g 50g
11497204 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11477204 Supplier Thermo Scientific Alfa Aesar Supplier No. B21594.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 76-37-9 | C3H4F4O | 132.06 g/mol

2,2,3,3-Tetrafluoro-1-propanol, is used as an essential fluorinated fine chemical, the main application of TFP is as a dye solvent in CD-R(W) and DVD-R(W) industry. It is also used in making textile treatment agent and pharmaceuticals. This chemical is an ideal replacement for Freon 22 detergent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3-Tetrafluoro-1-propanol, is used as an essential fluorinated fine chemical, the main application of TFP is as a dye solvent in CD-R(W) and DVD-R(W) industry. It is also used in making textile treatment agent and pharmaceuticals. This chemical is an ideal replacement for Freon 22 detergent.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-37-9
Molecular Formula C3H4F4O
Molecular Weight (g/mol) 132.06
MDL Number MFCD00004676
InChI Key NBUKAOOFKZFCGD-UHFFFAOYSA-N
Synonym 2,2,3,3-tetrafluoro-1-propanol, 2,2,3,3-tetrafluoropropanol, 1h,1h,3h-tetrafluoro-1-propanol, 1-propanol, 2,2,3,3-tetrafluoro, tetrafluoropropyl alcohol, 2,2,3,3-tetrafluoropropyl alcohol, 1,1,3-trihydroperfluoro-1-propanol, unii-rno513716p, 1h,1h,3h-tetrafluoropropanol, wln: q1xffyff
PubChem CID 6441
IUPAC Name 2,2,3,3-tetrafluoropropan-1-ol
SMILES OCC(F)(F)C(F)F

Specifications

Melting Point <-70°C
Density 1.471
Boiling Point 107°C to 109°C
Flash Point 49°C (120°F)
Refractive Index 1.3195
Quantity 10 g
UN Number UN1987
Beilstein 1738264
Merck Index 14,9206
Solubility Information Soluble in water.
Formula Weight 132.06
Percent Purity 97%
Chemical Name or Material 2,2,3,3-Tetrafluoro-1-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.