missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3-Tetrafluoro-1-propanol, 99+%

Product Code. 10617674 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10617674 100 g Glass bottle 100g
10401111 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10617674 Supplier Thermo Scientific Acros Supplier No. 296381000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 76-37-9 | C3H4F4O | 132.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-37-9
Molecular Formula C3H4F4O
Molecular Weight (g/mol) 132.06
MDL Number MFCD00004676
InChI Key NBUKAOOFKZFCGD-UHFFFAOYSA-N
Synonym 2,2,3,3-tetrafluoro-1-propanol, 2,2,3,3-tetrafluoropropanol, 1h,1h,3h-tetrafluoro-1-propanol, 1-propanol, 2,2,3,3-tetrafluoro, tetrafluoropropyl alcohol, 2,2,3,3-tetrafluoropropyl alcohol, 1,1,3-trihydroperfluoro-1-propanol, unii-rno513716p, 1h,1h,3h-tetrafluoropropanol, wln: q1xffyff
PubChem CID 6441
IUPAC Name 2,2,3,3-tetrafluoropropan-1-ol
SMILES OCC(F)(F)C(F)F

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless
Density 1.4700g/mL
Boiling Point 109°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula F2CHCF2CH2OH
Refractive Index 1.3200 to 1.322
Quantity 100 g
Merck Index 15, 9351
Specific Gravity 1.47
Formula Weight 132.06
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2,2,3,3-Tetrafluoro-1-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.