missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3-Tetrafluoro-1-propanol, TRC

Product Code. 30418969
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
30418969 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 30418969

Brand: Toronto Research Chemicals TRCT204733250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2,3,3-Tetrafluoro-1-propanol
CAS 76-37-9
Molecular Formula C3H4F4O
Quantity 250 mg
InChI Formula InChI=1S/C3H4F4O/c4-2(5)3(6,7)1-8/h2,8H,1H2
SMILES OCC(F)(F)C(F)F
IUPAC Name 2,2,3,3-tetrafluoropropan-1-ol
Molecular Weight (g/mol) 132.06
Formula Weight 132.0198
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.