missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,4,4,4-Hexafluoro-1-butanol, 95%

Product Code. 11498987
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498987 100 g 100g
11478987 5 g 5g
11488987 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498987 Supplier Thermo Scientific Alfa Aesar Supplier No. B20587.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 382-31-0 | C4H4F6O | 182.065 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 382-31-0
Molecular Formula C4H4F6O
Molecular Weight (g/mol) 182.065
MDL Number MFCD00042309
InChI Key LVFXLZRISXUAIL-UHFFFAOYSA-N
Synonym 2,2,3,4,4,4-hexafluoro-1-butanol, 1h,1h,3h-hexafluorobutanol, 2,2,3,4,4,4-hexafluorobutanol, 2,2,3,4,4,4-hexafluorobutanol-1, 2,2,3,4,4,4-hexafluoro-butan-1-ol, 1-butanol, 2,2,3,4,4,4-hexafluoro, 1h,1h,3h-perfluorobutan-1-ol, acmc-209iys, cf3cfhcf2ch2oh, 2,2,3,4,4,4-hexafluorobutyl alcohol
PubChem CID 533990
IUPAC Name 2,2,3,4,4,4-hexafluorobutan-1-ol
SMILES C(C(C(C(F)(F)F)F)(F)F)O

Specifications

Density 1.64
Boiling Point 114°C to 116°C
Flash Point 51°C (123°F)
Refractive Index 1.312
Quantity 100 g
UN Number UN1993
Beilstein 1753872
Formula Weight 182.07
Percent Purity 95%
Chemical Name or Material 2,2,3,4,4,4-Hexafluoro-1-butanol

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.