missing translation for 'onlineSavingsMsg'
Learn More

2-(2,3-dichlorophenyl)-1,3-thiazole-4-carboxylic acid, Thermo Scientific™

Product Code. 10382813
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
10382813 250 mg 250mg
10557563 1 g 1g
10653635 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 10382813

Brand: Thermo Scientific Maybridge SPB02181CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 257876-07-6
Molecular Formula C10H5Cl2NO2S
Molecular Weight (g/mol) 274.115
InChI Key ANTJCNIMRNBFMJ-UHFFFAOYSA-N
Synonym 2-2,3-dichlorophenyl-1,3-thiazole-4-carboxylic acid, 2-2,3-dichloro-phenyl-thiazole-4-carboxylic acid, 2-2,3-dichlorophenyl thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-2,3-dichlorophenyl
PubChem CID 2743987
IUPAC Name 2-(2,3-dichlorophenyl)-1,3-thiazole-4-carboxylic acid
SMILES C1=CC(=C(C(=C1)Cl)Cl)C2=NC(=CS2)C(=O)O

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 274.13
Percent Purity 97%
Chemical Name or Material 2-(2, 3-Dichlorophenyl)-1, 3-thiazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.