missing translation for 'onlineSavingsMsg'
Learn More

2-(2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethoxy)ethyl Acetate, TRC

Product Code. 30407465
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30407465 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407465 Supplier Toronto Research Chemicals Supplier No. TRCH94246525MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethoxy)ethyl Acetate
CAS 2518011-10-2
Molecular Formula C12H24N2O4
Quantity 25 mg
InChI Formula InChI=1S/C12H24N2O4/c1-12(16)18-11-10-17-9-7-14-4-2-13(3-5-14)6-8-15/h15H,2-11H2,1H3
SMILES CC(OCCOCCN1CCN(CCO)CC1)=O
IUPAC Name 2-(2-(4-(2-hydroxyethyl)piperazin-1-yl)ethoxy)ethyl acetate
Molecular Weight (g/mol) 260.33
Formula Weight 260.17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.